Learn More
Caplugs™ Evergreen Scientific FPC™ Fecal Parasite Concentrator Kit
Catalog No. 0973090 Shop All Caplugs Products
Change view
Click to view available options
Quantity:
100/Cs.
Catalog No. Quantity
09-730-90 100/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 09-730-90 Supplier Caplugs™ Supplier No. 3081000000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specimen collection kit; Fecal; Ova and parasite procedures; 100 tests; Includes preassembled centrifuge tube-strainer units, caps, dispensing spoons, 20% Triton X-100, trays, applicator sticks, biohazard bags, and instructions
Specifications
Product Type Fecal Parasite Concentrator Kit
Includes Preassembled centrifuge tube strainer units, Centrifuge tube caps, dispensing spoons, 20% Triton X-100, trays, applicator sticks, biohazard bags, largest filter matrix with 0.6 x 0.6mm holes and instructions
Quantity 100/Cs.