Learn More
Caplugs™ Evergreen Scientific FPC Fecal Parasite Concentrator Spoon
Catalog No. 055546 Shop All Caplugs Products
Change view
Click to view available options
Quantity:
100/Cs.
Catalog No. Quantity
055546 100/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 055546 Supplier Caplugs™ Supplier No. 2203596030
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safe and sanitary way to move helminth eggs and larvae, protozoan cysts, coccidian oocysts and microsporidian spores
  • For use with the FPC Fecal Parasite Concentrator Kit
  • 12.7cm (5 in.) long spoon
  • Made of polypropylene
  • Nonsterile
Specifications
Product Type Specimen Collection Spoon
Quantity 100/Cs.
Length (Metric) 5 in.