Learn More
Caplugs™ Evergreen Scientific Polystyrene Microplate, 96-Well
Catalog No. 055516 Shop All Caplugs Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
05-551-6 Clear
1 options
Catalog No. 05-551-6 Supplier Caplugs™ Supplier No. 290811701R
Add to Cart
Edge
Add to Cart
High quality polystyrene microplates
  • Wells feature raised rims to minimize cross-contamination
  • Optional fitting lids reduce evaporation
  • Precision molded for minimal variation
  • 96 wells
  • U-shape bottom
  • Non-sterile
Specifications
Lid Without Lid
Array 8 x 12
Color Clear
Product Type 96-well Assay Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Well Shape Round
Quantity 100/Pk.