Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caplugs™ Evergreen Scientific Specimen Containers

Catalog No. 055547 Shop All Caplugs Products
Change view
Click to view available options
Capacity (Metric):
118 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
055547 118 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 055547 Supplier Caplugs™ Supplier No. 2405614030
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sturdy enough to be reusable, but economical enough to be disposable

  • Useful for almost any storage need, such as in a laboratory or nursing station
  • Highly resistant to most chemicals, including formaldehyde, most bases and weak acids
  • Precision-molded from polypropylene
  • Can withstand boiling and freezing

Order Info

Available with or without caps

Specifications

Graduated Yes
Material Polyethylene
Capacity (English) 4 oz.
Capacity (Metric) 118 mL
Closure Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.