Learn More
Research Products International Corp Everlst Rcker 247 Sngle Pltfrm
SDP
Catalog No. 502979101
Encompass
The Everlast™ Rocker 247 provides the optimum agitation for blotting membranes or staining gels. Digital speed control accurately maintains set speed from 3 to 80 rpm. Produces a uniform and gentle wave pattern that washes over and under membranes. Adjustable stainless steel platform with locking tilt can be set from 0 to 20° arc. Exceptionally durable unit with powerful motor and heavy-duty drive system remains reliable and consistent. Operates from 0°C to +65°C and can be placed inside an incubator. Supplied with non-slip rubber mat. Five year warranty.
Catalog No. 50-297-9101 Supplier Research Products International Corp Supplier No. 400711
The Everlast™ Rocker 247 provides the optimum agitation for blotting membranes or staining gels. Digital speed control accurately maintains set speed from 3 to 80 rpm. Produces a uniform and gentle wave pattern that washes over and under membranes. Adjustable stainless steel platform with locking tilt can be set from 0 to 20° arc. Exceptionally durable unit with powerful motor and heavy-duty drive system remains reliable and consistent. Operates from 0°C to +65°C and can be placed inside an incubator. Supplied with non-slip rubber mat. Five year warranty.