Learn More
Cayman Chemical Evernic Acid, Purity: ≥98%, CAS: 537-09-7, 10 mg
SDP
Catalog No. 502980479
Encompass_Preferred
A secondary metabolite produced by some species of lichen that binds FAS-II enzymes to produce antibacterial and antiplasmodial effects; inhibits PfFabZ and PfFabI (IC50 = 10.7 and 36.1 µM, respectively) but shows low efficacy against the malaria parasite P. berghei (IC50 = 77.3 µM)
Catalog No. 50-298-0479 Supplier Cayman Chemical Supplier No. 1743010 MG
A secondary metabolite produced by some species of lichen that binds FAS-II enzymes to produce antibacterial and antiplasmodial effects; inhibits PfFabZ and PfFabI (IC50 = 10.7 and 36.1 µM, respectively) but shows low efficacy against the malaria parasite P. berghei (IC50 = 77.3 µM)