Learn More
Cayman Chemical EvernIc AcId 50mg
SDP
Catalog No. 502980553
Encompass_Preferred
A secondary metabolite produced by some species of lichen that binds FAS-II enzymes to produce antibacterial and antiplasmodial effects; inhibits PfFabZ and PfFabI (IC50 = 10.7 and 36.1 µM, respectively) but shows low efficacy against the malaria parasite P. berghei (IC50 = 77.3 µM)
Catalog No. 50-298-0553 Supplier Cayman Chemical Supplier No. 1743050 MG
A secondary metabolite produced by some species of lichen that binds FAS-II enzymes to produce antibacterial and antiplasmodial effects; inhibits PfFabZ and PfFabI (IC50 = 10.7 and 36.1 µM, respectively) but shows low efficacy against the malaria parasite P. berghei (IC50 = 77.3 µM)