Learn More
Medchemexpress LLC Evernic acid | 537-09-7 | MFCD00045824 | 99.7% | 332.30 g·mol^-1 | C17H16O7 | 5 MG
SDP
Catalog No. 5000264914
Encompass_Preferred
Evernic acid is a lichen-derived secondary metabolite provided as a high-purity small-molecule reagent for research and analytical applications. It has CAS 537-09-7, molecular formula C17H16O7, and molecular weight 332.30 g·mol⁻1. Typical uses include biochemical, pharmacological, and analytical studies.
  • High-purity compound (99.71%) suitable for analytical use.
  • Characterized chemical identity with CAS 537-09-7.
  • Confirmed molecular formula C17H16O7 and molecular weight 332.30 g·mol⁻1.
  • Supplied in small pack sizes for low-quantity experiments.
  • Soluble in DMSO, convenient for solution-based assays.
  • Intended for research use in biochemical and pharmacological studies.
Catalog No. 50-002-64914 Supplier Medchemexpress LLC Supplier No. HY1213625MG
Evernic acid is a lichen-derived secondary metabolite provided as a high-purity small-molecule reagent for research and analytical applications. It has CAS 537-09-7, molecular formula C17H16O7, and molecular weight 332.30 g·mol⁻1. Typical uses include biochemical, pharmacological, and analytical studies.
  • High-purity compound (99.71%) suitable for analytical use.
  • Characterized chemical identity with CAS 537-09-7.
  • Confirmed molecular formula C17H16O7 and molecular weight 332.30 g·mol⁻1.
  • Supplied in small pack sizes for low-quantity experiments.
  • Soluble in DMSO, convenient for solution-based assays.
  • Intended for research use in biochemical and pharmacological studies.