Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Evernic Acid, MP Biomedicals

Catalog No. ICN209482 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN209482 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN209482 Supplier MP Biomedicals, Inc Supplier No. 0520948280

Chemical Identifiers

CAS 537-09-7
Molecular Formula C17H16O7
Molecular Weight (g/mol) 332.308
InChI Key GODLCSLPZIBRMG-UHFFFAOYSA-N
Synonym evernic acid, unii-2eq5w5403j, lecanoric acid methyl ether, 2,6-cresotic acid, 4-methoxy-, 4-ester with 6-methyl-beta-resorcylate, 4-4-methoxy-6-methylsalicyloyl oxy-6-methylsalicylic acid, 2-hydroxy-4-2-hydroxy-4-methoxy-6-methylbenzoyloxy-6-methylbenzoic acid, benzoic acid, 2-hydroxy-4-2-hydroxy-4-methoxy-6-methylbenzoyl oxy-6-methyl, evernic acid, pruiss, evernsaure
PubChem CID 10829
IUPAC Name 2-hydroxy-4-(2-hydroxy-4-methoxy-6-methylbenzoyl)oxy-6-methylbenzoic acid
SMILES CC1=CC(=CC(=C1C(=O)OC2=CC(=C(C(=C2)C)C(=O)O)O)O)OC

Specifications

Quantity 100 mg
Formula Weight 332.3
Chemical Name or Material Evernic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.