Learn More
Medchemexpress LLC HY-10218 10mg , Everolimus CAS:159351-69-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1021810MG
Encompass_Preferred
HY-10218 10mg Everolimus CAS: 159351-69-6 Everolimus (RAD001) is a Rapamycin (HY-10219) derivative and a potent, selective and orally active mTOR1 inhibitor. Everolimus binds to FKBP-12 to generate an immunosuppressive complex. Everolimus inhibits tumor cells proliferation and induces cell apoptosis and autophagy. Everolimus has potent immunosuppressive and anticancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8972