Learn More
AdipoGen Everolimus
SDP
Catalog No. 502560971
Encompass_Preferred
Chemical. CAS 159351-69-6. Formula C53H83NO14. MW 958.2. Isolated from Streptomyces hygroscopicus. Macrolide antibiotic, inhibiting bacterial protein synthesis. Potent immunosuppressant. Binds with high affinity to the FK506 binding protein-12 FKBP-12 to generate an immunosuppressive complex that inhibits the activation of the mammalian target of rapamycin mTOR. More selective for the mTORC1 protein complex, with little impact on the mTORC2 complex, compared to Rapamycin. Anticancer agent. Inhibition of mTOR reduces the activity of effectors downstream, which leads to a blockage in the progression of cells from G1 into S phase, and subsequently inducing cell growth arrest, apoptosis and autophagy, resulting in reduction of cell proliferation, angiogenesis and glucose uptake. Inhibits tumor proliferation in vitro and in vivo.
Catalog No. 50-256-0971 Supplier AdipoGen Supplier No. CDXE0074M250
Chemical. CAS 159351-69-6. Formula C53H83NO14. MW 958.2. Isolated from Streptomyces hygroscopicus. Macrolide antibiotic, inhibiting bacterial protein synthesis. Potent immunosuppressant. Binds with high affinity to the FK506 binding protein-12 FKBP-12 to generate an immunosuppressive complex that inhibits the activation of the mammalian target of rapamycin mTOR. More selective for the mTORC1 protein complex, with little impact on the mTORC2 complex, compared to Rapamycin. Anticancer agent. Inhibition of mTOR reduces the activity of effectors downstream, which leads to a blockage in the progression of cells from G1 into S phase, and subsequently inducing cell growth arrest, apoptosis and autophagy, resulting in reduction of cell proliferation, angiogenesis and glucose uptake. Inhibits tumor proliferation in vitro and in vivo.