Learn More
Medchemexpress LLC HY-10218 50mg Medchemexpress, Everolimus CAS:159351-69-6 Purity:>98%
SDP
Catalog No. 502028701
Encompass_Preferred
Medchemexpress, HY-10218 50mg Everolimus CAS:159351-69-6 Everolimus (RAD001) is a Rapamycin derivative and a potent, selective and orally active mTOR1 inhibitor. Everolimus binds to FKBP-12 to generate an immunosuppressive complex. Everolimus inhibits tumor cells proliferation and induces cell apoptosis and autophagy. Everolimus has potent immunosuppressive and anticancer activities[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8701 Supplier Medchemexpress LLC Supplier No. HY1021850MG
Medchemexpress, HY-10218 50mg Everolimus CAS:159351-69-6 Everolimus (RAD001) is a Rapamycin derivative and a potent, selective and orally active mTOR1 inhibitor. Everolimus binds to FKBP-12 to generate an immunosuppressive complex. Everolimus inhibits tumor cells proliferation and induces cell apoptosis and autophagy. Everolimus has potent immunosuppressive and anticancer activities[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.