Learn More
MOLECULAR DEPOT LLC Everolimus BSA - 0.5 mg
Catalog No. 5000013584
Encompass_Preferred
Everolimus BSA Conjugate Solution is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to Everolimus A carbon spacer arm links BSA to the Everolimus moiety Each BSA molecule contains several Everolimus units Dilutions of the provided conjugate can be made with Phosphate Buffer Saline For in vitro research use only
Catalog No. 50-000-13584 Supplier MOLECULAR DEPOT LLC Supplier No. P2010024
Everolimus BSA Conjugate Solution is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to Everolimus A carbon spacer arm links BSA to the Everolimus moiety Each BSA molecule contains several Everolimus units Dilutions of the provided conjugate can be made with Phosphate Buffer Saline For in vitro research use only