Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Everolimus-d4 | 1338452-54-2 | MFCD21363580 | >99.0% | 962.25 | C53H79D4NO14 | 500 UG
SDP

Catalog No. 5000245914
Encompass_Preferred

Everolimus-d4 is the deuterium-labeled analog of everolimus provided as a stable isotope internal standard for analytical quantification. It retains the parent compound structure with four deuterium atoms and is intended for use in bioanalytical assays and method development.

  • Deuterium labeled internal standard (d4) for accurate quantitation.
  • High isotopic purity (>99% d1-d4), minimizing unlabeled impurity interference.
  • Suitable for LC-MS and GC-MS method development and routine analysis.
  • Molecular formula C53H79D4NO14; molecular weight ~962.25.
  • Supplied in preweighed analytical quantities; follow supplier storage recommendations.

Catalog No. 50-002-45914 Supplier Medchemexpress LLC Supplier No. HY10218S500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Everolimus-d4 is the deuterium-labeled analog of everolimus provided as a stable isotope internal standard for analytical quantification. It retains the parent compound structure with four deuterium atoms and is intended for use in bioanalytical assays and method development.

  • Deuterium labeled internal standard (d4) for accurate quantitation.
  • High isotopic purity (>99% d1-d4), minimizing unlabeled impurity interference.
  • Suitable for LC-MS and GC-MS method development and routine analysis.
  • Molecular formula C53H79D4NO14; molecular weight ~962.25.
  • Supplied in preweighed analytical quantities; follow supplier storage recommendations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.