Learn More
Abcam Everolimus, mTOR inhibitor, 5MG
Catalog No. 502706309
Encompass_Preferred
MW 958.2 Da, Purity >98%. Potent mTOR inhibitor (IC₅₀ = 1.6 - 2.4 nM). Immunosupressant and anticancer activity in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6309 Supplier Abcam Supplier No. AB1421515MG
MW 958.2 Da, Purity >98%. Potent mTOR inhibitor (IC₅₀ = 1.6 - 2.4 nM). Immunosupressant and anticancer activity in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement