Learn More
Selleck Chemical LLC Everolimus (RAD001) 10mg 159351-69-6
SDP
Catalog No. 508359
Encompass_Preferred
Everolimus (RAD001) is an mTOR inhibitor of FKBP12 with IC50 of 1.6-2.4 nM in a cell-free assay. Everolimus induces cell apoptosis and autophagy and inhibits tumor cells proliferation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-835-9 Supplier Selleck Chemical LLC Supplier No. S112010MG
Everolimus (RAD001) is an mTOR inhibitor of FKBP12 with IC50 of 1.6-2.4 nM in a cell-free assay. Everolimus induces cell apoptosis and autophagy and inhibits tumor cells proliferation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.