Learn More
System Biosciences LLC EVERY EV RNA ISOLATION KIT = 20 Preps
SDP
Catalog No. NC2313048
Encompass_Preferred
Obtain high yields of total EVery, including small RNAs, from already isolated Exosomes. The easy 30-min protocol delivers highly pure, highly concentrated RNA. EV samples vary drastically in their purity and concentration. Some EV samples from sera of diseased subjects and plasma bring up the challenge of heavy load of contaminating proteins that can potentially lead to column clogging thus lower yields. New column dimensions work just as smoothly for RNA isolation from all types of EV samples, irrespective of upstream EV isolation methods.
Catalog No. NC2313048 Supplier System Biosciences LLC Supplier No. EVERY100B1
Obtain high yields of total EVery, including small RNAs, from already isolated Exosomes. The easy 30-min protocol delivers highly pure, highly concentrated RNA. EV samples vary drastically in their purity and concentration. Some EV samples from sera of diseased subjects and plasma bring up the challenge of heavy load of contaminating proteins that can potentially lead to column clogging thus lower yields. New column dimensions work just as smoothly for RNA isolation from all types of EV samples, irrespective of upstream EV isolation methods.