Learn More
Bio Rad Laboratories EveryBlot Blocking Buffer, 500 mL
Catalog No. NC1798224
Encompass
EveryBlot Blocking Buffer provides 5 minute blocking and maximum sensitivity for all western blots regardless of detection method.
Catalog No. NC1798224 Supplier Bio Rad Laboratories Supplier No. 12010020
EveryBlot Blocking Buffer provides 5 minute blocking and maximum sensitivity for all western blots regardless of detection method.