Learn More
Novus Biologicals™ evGAG - Extracellular Vesicles Purification Kit

Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Target Species | Human |
| For Use With (Application) | Kit components are meant to run a total of 20 reactions. |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.