Learn More
Aviva Systems Biology Corp EVI1 Antibody - N-terminal region (ARP58384_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58384_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EVI1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EVI1. Alternate names: AML1-EVI-1, EVI-1, MDS1-EVI1, MGC163392, PRDM3, EVI1, MDS1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54079