Learn More
Description
- Self-sealing adhesive strip
- Write-on areas with chain of custody
- Preprinted control numbers on the bag and investigating officers receipt
- Puncture and tear resistant
- 4 mm polyethylene
- Made from durable, clear material for easy content visibility and equipped with a permanent self-sealing closure for secure storage.
Specifications
Specifications
| Dimensions (L x W) | 5 x 8 in. |
| Product Type | Evidence Bag |
| Color | Transparent with Blue Print |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
