Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Evidence I.D. Dots
SDPSCI_ED

Catalog No. S69036 Shop All Lynn Peavey Company Products
Change view
Click to view available options
Color:
Black, White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S69036 Black, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S69036 Supplier Lynn Peavey Company Supplier No. 5282
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For evidence documentation.

  • Peel and stick
  • Printed black on white
  • Diameter: 1 3/8 in.
  • Roll of 100 dots

Specifications

Color Black, White
Format Label
Product Type Evidence I.D Dots Label
For Use With (Application) For evidence documentation
Quantity 100/Pk.
Diameter (English) 1.37 in.
Diameter (Metric) 3.47 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.