Learn More
Mopec Evidence Marking Flags
SDP
Catalog No. 22446307 Shop All Mopec Products
Change view
Click to view available options
Color:
Pink
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-446-307 Yellow
22-446-294 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-446-307 Supplier Mopec Supplier No. FS440
Add to Cart
Add to Cart
Designed to clearly identify items at outdoor scenes before collection and documentation, these markers are mounted on wire staffs for convenient placement across different terrain. Printed yellow and unprinted pink options provide alternative ways to visibly mark evidence locations.
Construction
  • Wire staff easy to push into ground
Dimensions
  • 21 in. Long
Quantity
  • 100 per package
Options and Accessories
  • Pink and yellow flag options available
  • 'EVIDENCE - Do Not Touch' printed on yellow flag only (FS440)
Specifications
Color Yellow
Quantity 100/Pk.