Learn More
Mopec Evidence Seals
SDP
Catalog No. 22110522 Shop All Mopec Products
Change view
Click to view available options
Message:
Caution - Chemically Treated Evidence
Evidence : Warning/Police Seal
Evidence Seal: Exhibit Number
Evidence: Article Collected
Integrity Seal
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Message
22-110-522 Evidence Seal: Exhibit Number
22-110-466 Integrity Seal
22-110-494 Evidence : Warning/Police Seal
22-110-484 Evidence: Article Collected
22-110-485 Caution - Chemically Treated Evidence
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22-110-522 Supplier Mopec Supplier No. FT024
Add to Cart
Add to Cart
Pre-cut adhesive seals are designed to securely seal and identify evidence, with convenient dispensing for quick and easy application.
  • Available in multiple printed styles and color options
  • Pre-cut adhesive seal format
  • Permanent adhesive backing
  • Dispenser box with Pop-up tab design
  • Seals and identifies evidence containers
  • Provides strong, permanent adhesion
  • Easy to apply with the Pop-up dispenser design
  • Suitable for use on paper, plastic, and metal surfaces
Specifications
Dimensions (L x W) 3 x 1.5 in.
Product Type Evidence Seals
Quantity 100/Roll
Message Evidence Seal: Exhibit Number