Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Evixapodlin | 2374856-75-2 | 98.1% | C34H36Cl2N8O4 | 10MG
SDP

Catalog No. 5000261402
Encompass_Preferred

Evixapodlin is an orally bioavailable small-molecule inhibitor of programmed death-ligand 1 (PD-L1) used in research to modulate PD-1/PD-L1 protein-protein interactions. It has demonstrated potent activity in preclinical assays and has been evaluated in early clinical studies for advanced solid tumors.

  • Chemical name: Evixapodlin.
  • CAS number: 2374856-75-2.
  • Molecular formula: C34H36Cl2N8O4.
  • Molecular weight: 691.61.
  • Reported IC50: 0.213 nM (PD-1/PD-L1 protein-protein interaction).
  • Purity: 98.1% (HPLC).
  • Physical form: powder for research use.
  • Typical use: reagent for PD-1/PD-L1 pathway and oncology research.

Catalog No. 50-002-61402 Supplier Medchemexpress LLC Supplier No. HY13840710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Evixapodlin is an orally bioavailable small-molecule inhibitor of programmed death-ligand 1 (PD-L1) used in research to modulate PD-1/PD-L1 protein-protein interactions. It has demonstrated potent activity in preclinical assays and has been evaluated in early clinical studies for advanced solid tumors.

  • Chemical name: Evixapodlin.
  • CAS number: 2374856-75-2.
  • Molecular formula: C34H36Cl2N8O4.
  • Molecular weight: 691.61.
  • Reported IC50: 0.213 nM (PD-1/PD-L1 protein-protein interaction).
  • Purity: 98.1% (HPLC).
  • Physical form: powder for research use.
  • Typical use: reagent for PD-1/PD-L1 pathway and oncology research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.