Learn More
TARGETMOL CHEMICALS INC EVIXAPODLIN 5MG
SDP
Catalog No. 502580183
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Evixapodlin (PD-1/PD-L1-IN 7) is a human PD-1/PD-L1 protein/protein interaction inhibitor (IC50 0.213).Evixapodlin has anticancer and antiviral activities. purity: 99%
Catalog No. 50-258-0183 Supplier TARGETMOL CHEMICALS INC Supplier No. T364875MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Evixapodlin (PD-1/PD-L1-IN 7) is a human PD-1/PD-L1 protein/protein interaction inhibitor (IC50 0.213).Evixapodlin has anticancer and antiviral activities. purity: 99%