Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Evixapodlin (GS-4224) | 2374856-75-2 | 98.1% | 691.61 g·mol⁻¹ | C34H36Cl2N8O4 | 5 MG
SDP

Catalog No. 5000222150
Encompass_Preferred

Evixapodlin (GS-4224) is a potent small-molecule inhibitor of the PD-1/PD-L1 protein-protein interaction, reported with an IC50 of 0.213 nM and demonstrated anticancer and antiviral activity. It is supplied as a research-grade chemical for in vitro and preclinical studies.

  • High potency PD-1/PD-L1 PPI inhibition (IC50 ~0.213 nM).
  • Reported purity 98.1% by manufacturer.
  • Soluble in DMSO at 50 mg/mL for in vitro use.
  • Available in small milligram quantities and as DMSO solution for assay setup.
  • Recommended storage conditions to preserve stability of powder and solutions.
  • Suitable for biochemical and cellular immune checkpoint assays.

Catalog No. 50-002-22150 Supplier Medchemexpress LLC Supplier No. HY1384075MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Evixapodlin (GS-4224) is a potent small-molecule inhibitor of the PD-1/PD-L1 protein-protein interaction, reported with an IC50 of 0.213 nM and demonstrated anticancer and antiviral activity. It is supplied as a research-grade chemical for in vitro and preclinical studies.

  • High potency PD-1/PD-L1 PPI inhibition (IC50 ~0.213 nM).
  • Reported purity 98.1% by manufacturer.
  • Soluble in DMSO at 50 mg/mL for in vitro use.
  • Available in small milligram quantities and as DMSO solution for assay setup.
  • Recommended storage conditions to preserve stability of powder and solutions.
  • Suitable for biochemical and cellular immune checkpoint assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.