Learn More
Aviva Systems Biology Corp Evl Antibody - middle region (ARP40846_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40846_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Evl. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Mouse Evl. Alternate names: AI528774ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-900