Learn More
Cayman Chemical (+)-Evodiamine, Purity: ≥98%, CAS: 518-17-2, 50 mg
SDP
Catalog No. 502980265
Encompass_Preferred
A natural indole alkaloid with demonstrated beneficial effects in cancer, obesity, inflammation, and many other conditions; an AhR antagonist (Ki = 28 nM), activator of TRPV1 (EC50 = 45 nM), and inhibitor of signaling through NF-κB
Catalog No. 50-298-0265 Supplier Cayman Chemical Supplier No. 1688550 MG
A natural indole alkaloid with demonstrated beneficial effects in cancer, obesity, inflammation, and many other conditions; an AhR antagonist (Ki = 28 nM), activator of TRPV1 (EC50 = 45 nM), and inhibitor of signaling through NF-κB