Learn More
Medchemexpress LLC HY-N0114 50mg Medchemexpress, Evodiamine CAS:518-17-2 Purity:>98%
SDP
Catalog No. 501933402
Encompass_Preferred
Medchemexpress, HY-N0114 50mg Evodiamine CAS:518-17-2 Evodiamine is an alkaloid isolated from the fruit of Evodia rutaecarpa Bentham with diverse biological activities including anti-inflammatory, anti-obesity, and antitumor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3402 Supplier Medchemexpress LLC Supplier No. HYN011450MG
Medchemexpress, HY-N0114 50mg Evodiamine CAS:518-17-2 Evodiamine is an alkaloid isolated from the fruit of Evodia rutaecarpa Bentham with diverse biological activities including anti-inflammatory, anti-obesity, and antitumor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.