Learn More
Apexbio Technology LLC Evodiamine, 50mg Cas# 518-17-2
SDP
Supplier:  Apexbio Technology LLC N160050
Encompass_Preferred
Evodiamine is an indole alkaloid found in Evodia rutaescarpa. It acts as an anti-inflammatory compound; the mechanism of action may be inhibition of prostaglandin E2 synthesis or by cyclooxygenase 2 induction and NF-B activation.1 Evodiamine has been stu Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8567