Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Evodiamine, 98%

Catalog No. AC457890050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457890050 5 g
AC457890010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457890050 Supplier Thermo Scientific Chemicals Supplier No. 457890050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 518-17-2 | C19H17N3O | 303.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-17-2
Molecular Formula C19H17N3O
Molecular Weight (g/mol) 303.37
MDL Number MFCD06407824
InChI Key TXDUTHBFYKGSAH-SFHVURJKSA-N
Synonym evodiamine, +-evodiamine, unii-c01825bvnl, evodiamine, evodia rutaecarpa, d-evodiamine, evodiamine, +, s-14-methyl-8,13,13b,14-tetrahydroindolo 2',3':3,4 pyrido 2,1-b quinazolin-5 7h-one, pubchem18244, indolo 2',3':3,4 pyrido 2,1-b quinazolin-5 7h-one, 8,13,13b,14-tetrahydro-14-methyl-, 13bs, 13bs-14-methyl-8,13,13b,14-tetrahydroindolo 2',3':3,4 pyrido 2,1-b quinazolin-5 7h-one
PubChem CID 442088
ChEBI CHEBI:4948
IUPAC Name (1S)-21-methyl-3,13,21-triazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8,15,17,19-heptaen-14-one
SMILES CN1[C@H]2N(CCC3=C2NC2=CC=CC=C32)C(=O)C2=CC=CC=C12

Specifications

Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 303.36
Percent Purity 98%
Chemical Name or Material Evodiamine
Hazard Category Danger
Hazard Statement GHS H Statement Fatal if swallowed.
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.