Learn More
Selleck Chemical LLC Evodiamine 25mg 518-17-2 Isoevodiamine
SDP
Supplier:  Selleck Chemical LLC S238225MG
Encompass_Preferred
Evodiamine (Isoevodiamine), an alkaloid extract from the fruit of Evodiae Fructus exhibits antitumor activities against the human tumor cells. It is shown to inhibit NF-kB activation through suppression of IkB kinase activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-568-4