Learn More
Selleck Chemical LLC Evodiamine 100mg 518-17-2 Isoevodiamine
SDP
Catalog No. 505685
Encompass_Preferred
Evodiamine (Isoevodiamine), an alkaloid extract from the fruit of Evodiae Fructus exhibits antitumor activities against the human tumor cells. It is shown to inhibit NF-kB activation through suppression of IkB kinase activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-568-5 Supplier Selleck Chemical LLC Supplier No. S2382100MG
Evodiamine (Isoevodiamine), an alkaloid extract from the fruit of Evodiae Fructus exhibits antitumor activities against the human tumor cells. It is shown to inhibit NF-kB activation through suppression of IkB kinase activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.