Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-Evodiamine 98.0+%, TCI America™
SDP

Catalog No. E10121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10121G Supplier TCI America Supplier No. E10121G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 518-18-3
Molecular Formula C19H17N3O
Molecular Weight (g/mol) 303.37
MDL Number MFCD06407824
InChI Key TXDUTHBFYKGSAH-UHFFFAOYNA-N
Synonym 8,13,13b,14-Tetrahydro-14-methylindolo[2′C,3′C:3,4]pyrido[2,1-b]quinazolin-5(7H)-one
PubChem CID 151289
IUPAC Name 21-methyl-3,13,21-triazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8,15,17,19-heptaen-14-one
SMILES CN1C2N(CCC3=C2NC2=CC=CC=C32)C(=O)C2=CC=CC=C12

Specifications

Melting Point 272°C
Color White-Yellow
Quantity 1 g
Formula Weight 303.37
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-Evodiamine
RTECSNumber NM3454500
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.