Learn More
Medchemexpress LLC HY-10535 10mg , Evofosfamide CAS:918633-87-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1053510MG
Encompass_Preferred
HY-10535 10mg Evofosfamide CAS: 918633-87-1 Evofosfamide (TH-302) is a hypoxia-activated prodrug with IC50 of 10 μM and 1000 μM in hypoxia (N2) and normoxia (21% O2), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9918