Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Evogliptin (tartrate) | 1222102-51-3 | 99.7% | 551.51 g/mol | C23H32F3N3O9 | 1 ML
SDP

Catalog No. 5000226614
Encompass_Preferred

Evogliptin tartrate is the tartrate salt of evogliptin (DA-1229), a potent and selective dipeptidyl peptidase-4 (DPP-4) inhibitor supplied for research use. Supplied as a 10 mM solution in DMSO (1 mL), it is used in in vitro and preclinical studies of glucose metabolism and related pathways. Consult the product datasheet and SDS for handling, storage, and safety information.

  • Potent, selective DPP-4 inhibitor for metabolic research.
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • High purity reported (~99.7%), suitable for analytical applications.
  • Molecular weight 551.51 g/mol; formula C23H32F3N3O9.
  • Intended for research use; follow the SDS for safe handling and storage.

Catalog No. 50-002-26614 Supplier Medchemexpress LLC Supplier No. HY117985B10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Evogliptin tartrate is the tartrate salt of evogliptin (DA-1229), a potent and selective dipeptidyl peptidase-4 (DPP-4) inhibitor supplied for research use. Supplied as a 10 mM solution in DMSO (1 mL), it is used in in vitro and preclinical studies of glucose metabolism and related pathways. Consult the product datasheet and SDS for handling, storage, and safety information.

  • Potent, selective DPP-4 inhibitor for metabolic research.
  • Supplied as a 10 mM solution in DMSO, 1 mL volume.
  • High purity reported (~99.7%), suitable for analytical applications.
  • Molecular weight 551.51 g/mol; formula C23H32F3N3O9.
  • Intended for research use; follow the SDS for safe handling and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.