Learn More
Trajan Scientific EVOL MEPS SYRINGE
SDP
Catalog No. 502530069
Encompass_Preferred
SGE MEPS Syringes, Volume: 500 uL, For Use With: eVol automated syringe, Gas/Liquid chromatography, Quantity: Each
Catalog No. 50-253-0069 Supplier Trajan Scientific Supplier No. 2910026
SGE MEPS Syringes, Volume: 500 uL, For Use With: eVol automated syringe, Gas/Liquid chromatography, Quantity: Each