Learn More
Medchemexpress LLC HY-P9930 1mg , Evolocumab CAS:1256937-27-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99301MG
Encompass_Preferred
HY-P9930 1mg Evolocumab CAS: 1256937-27-5 Evolocumab (AMG 145) is a human monoclonal antibody that inhibits PCSK9. Evolocumab binds to the circulating PCSK9 protein, inhibiting it from binding to the LDLR. Evolocumab can be used for the research of hypercholesterolemia and atherosclerotic cardiovascular diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1591