Learn More
HEPATOCHEM EvoluChem LED PF series LED 275 nm for photochemistry Dimmable 110V
Catalog No. NC3734518
Encompass_Preferred
The EvoluChem™ LED spotlights are designed specifically for photocatalytic, photochemistry or photobiology applications. This LED fits the PhotoRedOx Box™ UV/Vis HCK1022-01-001 only. Note that LEDs lower than 365 nm work only with the PhotoRedOx Box™ UV/Vis. They are designed to irradiate all samples with maximum efficiency compared to higher power LED light sources from the competition. They are more than 20 specific wavelengths ranging from 254 nm to 808 nm. This LED light wavelength is 275 nm. The beam angle is 30 Degrees. The intensity can be controlled by an external dimmer or by the PhotoRedOx Box™UV/Vis HCK1022-01-001. You may need to order a power cord HCK1006-01-034
Catalog No. NC3734518 Supplier HEPATOCHEM Supplier No. HCK101204026
The EvoluChem™ LED spotlights are designed specifically for photocatalytic, photochemistry or photobiology applications. This LED fits the PhotoRedOx Box™ UV/Vis HCK1022-01-001 only. Note that LEDs lower than 365 nm work only with the PhotoRedOx Box™ UV/Vis. They are designed to irradiate all samples with maximum efficiency compared to higher power LED light sources from the competition. They are more than 20 specific wavelengths ranging from 254 nm to 808 nm. This LED light wavelength is 275 nm. The beam angle is 30 Degrees. The intensity can be controlled by an external dimmer or by the PhotoRedOx Box™UV/Vis HCK1022-01-001. You may need to order a power cord HCK1006-01-034