Description
Thermo Scientific™ Mercury Lamp accessories provides instrument calibration and automated performance testing for the Thermo Scientific™ Evolution™ Series UV-Visible spectrophotometers. Test performance of wavelength accuracy, wavelength repeatability and bandwidth accuracy using the extremely narrow emission lines of mercury. These emission lines are fundamental, primary standards that require no certification or recalibration, saving you time and money.
- Calibrate the Evolution 200 Series or Evolution 300 spectrophotometer exactly as it was in the factory
- Uses seven mercury lamp lines from wavelength 253.65 to 810nm
- A unique calibration curve is used for every spectral bandwidth (SBW) available with the spectrophotometer Calibration at 1.0nm SBW is performed and stored independently of the calibration at 2.0nm SBW
- Features kinematic mounting for easy installation, allowing precise lamp alignment in the optical path
- Lamp can remain in place during normal Evolution 300 operation.
Specifications
Specifications
| Type | Lamp Calibration Accessory |
| For Use With (Equipment) | Evolution One/One Plus Spec |