Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Evolution™ One UV-Vis Spectrophotometer Loyalty Discount

Catalog No. 14385500LPM
Change view
Click to view available options
Detector Type:
Dual Silicon Photodiodes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detector Type
14-385-500LPM Dual Silicon Photodiodes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-385-500LPM Supplier Thermo Scientific™ Supplier No. 8403414001LD22
Add to Cart
Add to Cart

Thermo Scientific™ Evolution™ UV-Vis Spectrophotometers combine reliable and versatile hardware with easy-to-learn, easy-to-use Thermo Scientific™ Insight™ Pro Software. Together they will meet the diverse needs your organization requires from an analytical tool.

For routine to research analysis, this instrument series offers real innovations that provide enhanced usability and performance - without added complexity. With straightforward software, cutting-edge instrumentation and a wide selection of accessories, the Evolution One and One Plus UV-Vis Spectrophometers work with you to perform experiments the way you want - for the results you need.

Optimized Engineering Delivers Ultimate Performance:

  • Double-beam geometry with fixed 1 nm or variable spectral bandwidth options
  • Application Focused Beam Geometry (AFBG) technology enables the best performance from microcells, solid samples, and fiber optic probes (Evolution One Plus Only)
  • Xenon flash lamp requires no warm-up time allowing instant measurements
  • Meet chemical, materials science, or biological sampling and measurement needs with a broad range of accessories

Insight Pro Software—Easy-to-learn, easy-to-use

  • Reduce training time: Optimized user interface with prompting software defines clear step-by-step workflow and is easy to teach and learn
  • Prevent costly sampling and measurement errors: Create dedicated workflows for complex assay and transform multi-step workflows into one simplified, easy-to-follow method suitable for any level of user with Customized User Environment (CUE)
  • Always current: Whether it is user inputs or application requirements, or new regulations, Insight Pro Software is continuously improving with quick upgrades to deliver greater capabilities and efficiencies for your lab’s needs

Accessories for all your Sampling Needs:

  • Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete your analysis
  • Choice of Peltier Single Cell Holder or a Smart 8-Cell Peltier system for temperature control and sample monitoring from 0 to 100°C
  • Easy software integration gives you complete control of your measurements

Regulatory/Quality Control Environments:
To use the EvolutionOne/One Plus UV-Vis Spectrophotometers in a regulated or quality control environment, we offer a wide selection of compliance products and services to reduce implementation time and assure system performance.

Evolution One/One Plus Security and Validator Bundles include:

  • Evolution One or One Plus UV-Vis Spectrometer
  • INSIGHT Pro with Security Software
  • Validator for Insight Pro US Ltr or INTL A4 Paper Version
  • Qualification Filter Kit for UV-Vis

Evolution One/One Plus USP/EP Standards Pharma Bundles include:

  • Evolution One or One Plus Spectrometer
  • INSIGHT Pro with Security Software
  • Validator for Insight Pro US Ltr or INTL A4 Paper Version
  • USP and EP UV Standards Set
  • USP and EP Visible Standards Set
TRUSTED_SUSTAINABILITY

Specifications

Baseline Flatness ±0.001 A, 200 to 800 nm, 1.0 nm SBW, smoothed
Data Interval 10, 5, 2, 1.0, 0.5, 0.2, 0.1 nm
Description Evolution One UV-Vis Spectrophotometer
Detector Type Dual Silicon Photodiodes
Dimensions (L x W x H) 593 x 475 x 266 mm (23.3 x 18.7 x 10.6 in.)
Light Source Xenon Flash Lamp (>5 years typical, 3 years guaranteed)
Spectral Bandwidth 1 nm
Wavelength Range 190 to 1100 nm
Drift <0.0005 A/hr; 500 nm, 1.0 nm SBW, 1 hour warm-up
Electrical Requirements 100 to 240 V; 50 to 60 Hz, selected automatically, 150 W max.
Noise 0A: ≤0.00015 A; 1A: ≤0.00025 A; 2A: ≤0.00050 A; 260 nm, 1.0 nm SBW, RMS
Optical Design Double beam with sample and reference cuvette positions Czerny-Turner Monochromator
Pharmacopoeia Compliance Testing Photometric Accuracy (60 mg/L K2Cr2O7): ±0.010 A; Stray Light: ≤1%T at 198 nm: KCI; ≤0.05%AT at 220 nm: Nal, Kl; Wavelength Accuracy: ±.5 nm 541.9, 546.1 nm Hg emission lines, ±0.8 nm full range; Wavelength Repeatability: ≤0.05 nm, repetitive scanning of 546.1 nm Hg emission line
Photometric Accuracy Instrument 1A: ±0.004 A; 2A: ±0.008 A; Measured at 440 nm using neutral density filters traceable to NIST
Photometric Display -0.3 to 4.0 A
Photometric Range >3.5 A
Photometric Repeatability 1 A: ±0.0002 A
Scan Ordinate Modes Absorbance, % Transmittance, % Reflectance, Kubelka-Munk, log (1/R), log (Abs), Abs Factor, Intensity
Scan Speed <1 to 6000 nm/min, variable
Wattage 150 W max.
Wavelength Accuracy ±0.5 nm (541.9, 546.1 nm mercury lines); ±0.8 nm (full range 190 to 1100 nm)
Wavelength Repeatability ≤0.05 nm (546.1 nm mercury line, SD of 10 measurements)
Weight (English) 32 lb.
Weight (Metric) 14.5 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.