Learn More
Thermo Scientific™ Evolution UV-Vis Spectrophotometer Peltier Bundles
Catalog No. 14380503 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
E-Pro with 6-Cell Peltier
Catalog No. Type
14-380-503 E-Pro with 6-Cell Peltier
1 options
Catalog No. 14-380-503 Supplier Thermo Scientific™ Supplier No. 912A1283
Add to Cart
Add to Cart
Request a Quote
Achieve superior performance in temperature control and sample stirring for many UV-Visible analysis methods.
Thermo Scientific™ Peltier Accessories for the Thermo Scientific™ Evolution™ One, Evolution™ One Plus and Evolution™ Pro UV-Vis Spectrophotometers provide thermostatting and ramping with precise temperature control which is important for many UV-Visible analysis methods. Outperforming traditional heater/chiller units, these accessories offer thermostatting and ramping with precise temperature control between 0 °C and 110 °C. Add superior performance with compact integrated design, simple set-up, and fast, precise temperature control.
Precise Thermostatting
Peltier Accessories for the Evolution UV-Visible Spectrophotometers utilize re-circulating water as an effective heat sink to allow the Peltier element to heat or cool the block over a wide temperature range. The single-cell holder can reach as low as 0°C and as high at 110°C. Thermo Scientific offers 3 Peltier options to cover many possible applications, including a multi-cell unit. Stirring control for the single cell holders is provided when you place a micro-stir bar inside the cuvette. Unlike thermostatting with a traditional re-circulating water bath where a large volume of liquid must be heated or cooled, heating or cooling the cell holder with the Peltier effect means more rapid and more precise temperature control. Choose between water or chemical coolant to best fit your needs.
Temperature Ramping
Thermo Scientific INSIGHT Pro Software allows you to control and monitor the block temperature of the Peltier unit and allows you to implement programmed temperature ramping with multiple ramp and hold cycles and variable temperature kinetics.  Add a temperature probe hub and probes, and you can monitor the exact temperature in each cell as you perform heating and cooling cycles. The Insight Pro software also includes pre-set methods that can be used to study DNA denaturation and renaturation or other complex temperature programmed experiments.
TRUSTED_SUSTAINABILITY
Specifications
Type E-Pro with 6-Cell Peltier
Disposable Reusable
For Use With (Equipment) Evolution Pro with 6-Cell Rotary Peltier
Includes Kit consists of: Peltier cell holder, Temperature controller, Water tank and motor for cooling, All necessary tubing, cables, and connectors, Evolution Pro UV-Vis Spectrophotometer