Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlington Scientific Evolution RPR Titration Diluent
SDP

Catalog No. 22415221 Shop All Arlington Scientific Products
Change view
Click to view available options
Clia Complexity:
Moderate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
22-415-221 Moderate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-415-221 Supplier Arlington Scientific Supplier No. 5300905A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in nontreponemal tests for syphilis.

  • Assists in establishing reactivity patterns for samples
  • Titer function can be used in the evaluation of the reactivity of nontreponemal test antigens

Specifications

Content And Storage 2°C to 8°C
CPT Code 86592
Description Sample Diluent
CE Marker Self-declared
Clia Complexity Moderate
Packaging Type Bottle
Quantity 20 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.