Learn More
World Precision Instrument EVOM™ AUTOLCI, Brightfield, 4X Magnification, with Green Fluorescence
SDP
Supplier:  World Precision Instrument EVILCI0103
Encompass
EVOM™ AutoLCI is an automated live cell imaging system that is equipped with an advanced fluorescence and bright field microscopy, auto-focusing and real-time, multi-position imaging technology for a well plate, dish or T-flask. The streamlined process provides an easy workflow solution giving you a full set of tools you need to acquire the best quality images and accurate research results. The compact nature of the EVOM™ AutoLCI allows positioning in an incubator, providing improved cell viability, because there are fewer disturbances over the course of your experiment, reducing chances of cellular abnormality.
Catalog No. 50-252-1189
Only null left
Add to Cart
Request a Quote