Learn More
World Precision Instrument EVOM™ Manual Meter for TEER Measurement
SDP
Catalog No. NC3589687
Encompass
The EVOM™ Manual replaces all previously manufactured WPI manual TEER meters, including the EVOM3, EVOM2, and MilliCell ERS-2, which have now been discontinued.This next-generation manual TEER meter features automatic data logging for streamlined data collection. Its low-noise design ensures greater resolution and accuracy, while the automatic 20X sample averaging enhances measurement stability and precision. Select from adjustable fixed measurement currents (2, 4, or 10 μA) to suit your protocol. The auto-ranging resistance measurements span from 1 Ω to 100,000 Ω, with three fixed current range options for added flexibility.
Enjoy continuous data logging via USB flash drive, which is compatible with PC, Mac, and Linux.
NEW FEATURE: In addition to the existing capability of data storage on a USB flash drive, the new version of the EVOM™ Manual now provides an option for a more secured mode of data transfer using a Windows companion application.
Catalog No. NC3589687 Supplier World Precision Instrument Supplier No. EVMMT0302
The EVOM™ Manual replaces all previously manufactured WPI manual TEER meters, including the EVOM3, EVOM2, and MilliCell ERS-2, which have now been discontinued.This next-generation manual TEER meter features automatic data logging for streamlined data collection. Its low-noise design ensures greater resolution and accuracy, while the automatic 20X sample averaging enhances measurement stability and precision. Select from adjustable fixed measurement currents (2, 4, or 10 μA) to suit your protocol. The auto-ranging resistance measurements span from 1 Ω to 100,000 Ω, with three fixed current range options for added flexibility.
Enjoy continuous data logging via USB flash drive, which is compatible with PC, Mac, and Linux.
NEW FEATURE: In addition to the existing capability of data storage on a USB flash drive, the new version of the EVOM™ Manual now provides an option for a more secured mode of data transfer using a Windows companion application.