Learn More
World Precision Instrument EVOM STX Calibration Solution
SDP
Catalog No. 502538860
Encompass
  • Calibration solution for your TEER measuring system electrodes.
  • This can be used for troubleshooting any issues you are having with electrodes.
  • Volume of 125 mL
  • Package of 5
Catalog No. 50-253-8860 Supplier World Precision Instrument Supplier No. EVOMCAL
  • Calibration solution for your TEER measuring system electrodes.
  • This can be used for troubleshooting any issues you are having with electrodes.
  • Volume of 125 mL
  • Package of 5