Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EVOS™ Light Cube, CFP-YFP EM 2.0

Catalog No. 12563692
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-563-692 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-563-692 Supplier Invitrogen™ Supplier No. AMEP4969
  • $5,605.00
    $4,195.00 / Each of 1

    Save $1,410.00

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality. At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through.

  • Second-generation EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality
  • At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through
  • Like the original design, they combine condensing optics and hard-coated filters into one light engine that delivers digitally controlled fluorophore excitation
  • These LED light cubes are backwards compatible with older EVOS and Countess systems. and can be used with first-generation light cubes
  • Like the original light cubes, these LED cubes are user-changeable and automatically recognized by the instrument
  • The EVOS Light Cube, CFP-YFP EM 2.0, is ideal for use with CFP-YFP FRET emission, and other fluorophores with excitation and emission maxima near 445 and 542 nm
  • The LED bulbs on the EVOS systems are rated for >50,000 hours, compared to 300 hours for a typical mercury bulb and 1,500 hours for a metal halide bulb

EVOS LED light cubes offer these important advantages:

  • Uniform illumination across the entire field of view for superb image quality
  • Optimized spectral compatibility to maximize true signal detection, while minimizing bleed-through for the best possible data quality
  • A short light path that affords superior detection of fluorescent signals
  • Stable illumination intensity over 50,000 hours that helps experimental reproducibility
TRUSTED_SUSTAINABILITY

Specifications

Type LED Light Cube
Product Line EVOS
Excitation/Emission Ex: 445/45 nm Em: 542/27
For Use With (Equipment) Countess™ III FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System
Includes Condensing Optics, Hard-coated Filters
Quantity 1 each

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.