Learn More
Invitrogen™ EVOS™ Light Cube, QDOT WB 525-800 2.0
Catalog No. 12563695
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
12-563-695 1 each
1 options
Catalog No. 12-563-695 Supplier Invitrogen™ Supplier No. AMEP4966
  • $5,959.00
    $4,213.00 / Each of 1
    Save $1,746.00
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote
EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality. At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through.
  • Second-generation EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality
  • At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through
  • Like the original design, they combine condensing optics and hard-coated filters into one light engine that delivers digitally controlled fluorophore excitation
  • These LED light cubes are backwards compatible with older EVOS and Countess systems. and can be used with first-generation light cubes
  • Like the original light cubes, these LED cubes are user-changeable and automatically recognized by the instrument
  • The EVOS Light Cube, QDOT WB 525-800 2.0, is ideal for use with Qdot fluorophores, and other fluorophores with excitation and emission maxima near 445 and 514 nm
  • The LED bulbs on the EVOS systems are rated for >50,000 hours, compared to 300 hours for a typical mercury bulb and 1,500 hours for a metal halide bulb
EVOS LED light cubes offer these important advantages:
  • Uniform illumination across the entire field of view for superb image quality
  • Optimized spectral compatibility to maximize true signal detection, while minimizing bleed-through for the best possible data quality
  • A short light path that affords superior detection of fluorescent signals
  • Stable illumination intensity over 50,000 hours that helps experimental reproducibility
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 each
Can EVOS Light Cube, QDOT WB 525-800 2.0 (Cat. No. AMEP4966), be used to image other dyes, besides Qdots?
The ability to use EVOS Light Cube, QDOT WB 525-800 2.0 (Cat. No. AMEP4966) to image other dyes besides Qdots depends on the excitation/emission characteristics of the dye. QDOT cubes are excited at 445 nm and have a 514 nm long-pass filter. If the dye fits within these spectral characteristics, the dye can be imaged with the EVOS Light Cube, QDOT WB 525-800 2.0 (Cat. No. AMEP4966). However, other EVOS Light Cubes (e.g., GFP, RFP, etc.) are usually more suitable.
Are EVOS 2.0 Light Cubes compatible with the Countess II FL?
Yes. All our EVOS Light Cubes can be used with the Countess II FL.
Are EVOS 2.0 Light Cubes compatible with older EVOS instruments?
Yes. Our EVOS 2.0 Light cubes can be used in all EVOS instruments with EVOS light cube functionality.
For Research Use Only. Not for use in diagnostic procedures.