Learn More
Thermo Scientific™ EVOS™ Vessel Plate for Large Vessels
Description
This EVOS vessel plate is intended for use on the stage of the EVOS FL Color Imaging System. This plate is designed to hold large vessels, for increased precision in sample alignment and is just one of many easy-to-change plates and holders available for this instrument platform. With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
Specifications
Specifications
| Type | Vessel Plate |
| Product Line | EVOS |
| Device Holds | Large Vessel |
| For Use With (Equipment) | EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Quantity | 1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.